Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCND1 Mouse Monoclonal Antibody [clone: 5D4]

Supplier: Enzo Life Sciences
KAM-CC200-E
10050-330EA 365.35 USD InStock
10050-330
Anti-CCND1 Mouse Monoclonal Antibody [clone: 5D4]
Antibodies
Cyclins are a family of proteins which function as the regulatory subunits of cyclin/cyclin-dependent kinase (Cdk) holoenzymes, mediating entry into and progression through the cell cycle. Each cyclin is expressed in a temporal fashion during the cell cycle, partnering with a specific Cdk to activate its protein kinase activity. The most well defined mitotic cyclins are: cyclin D, which partners with cdks 4 and 6 during early G1 phase; cyclin E, which activates Cdk2 in the G1 to S phase transition; cyclin A, which utilizes Cdks 1 and 2 during progression through S phase and into G2; and cyclin B, which activates Cdk1 in M-phase promotion.

Type: Primary
Antigen: CCND1
Clonality: Monoclonal
Clone: 5D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty