Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TOP2A Mouse Monoclonal Antibody [clone: 1C5]

Supplier: Enzo Life Sciences
KAM-CC210-E
10050-332EA 460.64 USD InStock
10050-332
Anti-TOP2A Mouse Monoclonal Antibody [clone: 1C5]
Antibodies
DNA-topoisomerase II, also known as Topo II or Top 2, is a multifunctional nuclear enzyme required during DNA replication, chromosome disjunction at mitosis, and other DNA-related activities because of its ability to alter DNA supercoiling. In mammalian cells, Topo II consists of two isozymes, Topo II-alpha (170kDa) and Topo II-beta (180kDa) and expression and localization of each isoform are distinct and stage specific during the cell cycle. Topo II-beta is expressed constitutively throughout the cell cycle, whereas the expression of Topo II-alpha is cell cycle-regulated, peaking in G2 to M phase and declining to a minimal level at the end of M phase. DNA-topoisomerase II is the target of some of the most active drugs used in chemotherapy and has been shown to have a critical role in neural development.

Type: Primary
Antigen: TOP2A
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty