Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PI3 Kinase Mouse Monoclonal Antibody [clone: AB6]

Supplier: Enzo Life Sciences
KAM-PI200-E
10050-138EA 399.46 USD InStock
10050-138
Anti-PI3 Kinase Mouse Monoclonal Antibody [clone: AB6]
Antibodies
Phosphatidylinositol 3-kinase (PI3-K) is a lipid kinase that phosphorylates the inositol ring of phosphatidylinositol and related compounds at the 3

Type: Primary
Antigen: PI3 kinase
Clonality: Monoclonal
Clone: AB6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty