Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SHPS-1/SIRP-1 alpha Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-TK140-E
10049-388EA 513 USD InStock
10049-388
Anti-SHPS-1/SIRP-1 alpha Rabbit Polyclonal Antibody
Antibodies
SHPS-1 is a member of the gene family called the signal regulatory proteins of which there are at least fifteen members. SHPS-1 is a substrate of many activated tyrosine kinases such as Insulin receptor and EGFR, amongst others. SHPS-1 has regulatory effects on cellular responses induced by serum, growth factors, insulin, oncogenes, growth hormones and cell adhesion and plays a general role in different physiological and pathological processes.

Type: Primary
Antigen: SHPS-1/SIRP-1 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-Terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty