Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Vesicular stomatitis virus tag Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-MSA-115-E
10049-406EA 365.35 USD InStock
10049-406
Anti-Vesicular stomatitis virus tag Rabbit Polyclonal Antibody
Antibodies
Epitope tagging is a widely accepted technique that fuses an epitope peptide to a certain protein as a marker for gene expression. With this technique, gene expression can be easily monitored on Western blotting, immunoprecipitation and immunofluorescence utilizing an antibody that recognizes such an epitope. The VSV-G Tag (YTDIEMNRLGK) corresponds to the partial peptide sequence of the vesicular stomatitis virus glycoprotein.

Type: Tag
Antigen: Vesicular stomatitis virus tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty