Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-p47Phox, NCF1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
OSA-550-0050
10049-440EA 237.99 USD InStock
10049-440
Anti-p47Phox, NCF1 Rabbit Polyclonal Antibody
Antibodies
Originally recognized for their role in the respiratory burst of phagocytes, the NOX family of NADPH oxidases is now known to consist of at least 13 enzymes and enzyme subunits that catalyze the production of superoxide and other downstream reactive oxygen species (ROS) in nearly all cells and tissues. NOX enzymes contribute to a multitude of physiological functions including host defense, cell signaling, and differentiation, with dysregulation linked to pathological manifestations including immunosuppression, hypothyroidism, cardiovascular disease, and neurodegeneration. p47Phox, also known as neutrophil cytosolic factor 1 (NCF1), is considered the organizing subunit of NADPH oxidase enzyme complexes containing the prototypic NOX2 enzyme. Cell stimulation results in phosphorylation of p47Phox, exposing a proline rich region which mediates translocation and association with membrane-associated NOX2/p22 complexes. Translocation of p47Phox is accompanied by the p40Phox subunit and p67Phox activator subunit, which along with GTP-bound Rac completes formation of the active NOX2 oxidase complex. Defects in p47Phox are associated with chronic granulomatous disease (CGD), characterized by an inability of host macrophages and phagocytes to kill ingested microbes, leading to life-threatening infections.

Type: Primary
Antigen: p47Phox, NCF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty