Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FLAG Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SAB-410-0100
10049-460EA 311.82 USD InStock
10049-460
Anti-FLAG Rabbit Polyclonal Antibody
Antibodies
FLAG-tag stands for an octapeptide with a sequence of DYKDDDDK. It is a polypeptide protein tag that can be added to a protein using recombinant DNA technology. It is widely used for affinity chromatography to purify recombinant proteins from variety of systems. It can also be used in the isolation of protein complexes with multiple subunits. A FLAG-tag can be used in many different assays that require recognition by an antibody. If there is no antibody against the studied protein, adding a FLAG-tag to this protein allows one to follow the protein with an antibody against the FLAG sequence, such as in ELISA, Western Blot, etc.

Type: Primary
Antigen: FLAG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Species independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty