Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRYAB Mouse Monoclonal Antibody [clone: 1B6.1-3G4]

Supplier: Enzo Life Sciences
ADI-SPA-222-D
10049-488EA 282.08 USD InStock
10049-488
Anti-CRYAB Mouse Monoclonal Antibody [clone: 1B6.1-3G4]
Antibodies
Alpha-crystallins, which are part of the small Heat shock family members, are major water-soluble proteins present in the lens of the mammalian eye. Phosphorylation of serine residues which occurs during development and in response to stress, is intimately linked with its function. Chaperone activity requires, and is modulated by, oligomerization and is limited to binding unfolded intermediates to prevent irreversible aggregation.

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 1B6.1-3G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty