Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRPE Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-240-J
95042-930EA 2053.82 USD InStock
95042-930
Anti-GRPE Rabbit Polyclonal Antibody
Antibodies
GrpE (Glucose Regulated Protein E. coli) is encoded by a monocistronic E. coli gene, which is under the control of both s32 and s70 transcription factors. The GrpE protein is a so-called co-chaperone because it is known to assist the DnaK (Hsp70) protein to effectively carry out DnaK-dependent chaperone activity (i.e. protein folding, protein transport, disaggregation of heat inactivated proteins, activation of mutant protein).

Type: Primary
Antigen: GrpE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty