Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP70 Mouse Monoclonal Antibody (DyLight® 488) [clone: C92F3A-5]

Supplier: Enzo Life Sciences
SPA-810-488-D
95042-192EA 300.82 USD InStock
95042-192
Anti-HSP70 Mouse Monoclonal Antibody (DyLight® 488) [clone: C92F3A-5]
Antibodies
Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone: C92F3A-5
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Fish, Drosophila, Dog, Chicken, C. elegans, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty