Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP90B1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-851-D
95042-134EA 242.56 USD InStock
95042-134
Anti-HSP90B1 Rabbit Polyclonal Antibody
Antibodies
Grp94 (Glucose-regulated protein 94) is an abundant resident endoplasmic reticulum (ER) lumenal stress protein, which together with cytosolic Hsp90 belongs to the Hsp90 family of molecular chaperones. Grp94 expression is upregulated by stress conditions such as glucose starvation and heat shock, which promote protein misfolding or unfolding. In addition to a homeostatic role in protein folding and assembly, Grp94 can function in the intracellular trafficking of peptides from the extracellular space to the MHC class I antigen processing pathway of antigen presentation cells.

Type: Primary
Antigen: HSP90B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-Terminus
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty