Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACY1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11316-1-AP-150UL
10272-976EA 453 USD InStock
10272-976
Anti-ACY1 Rabbit Polyclonal Antibody
Antibodies
The Aminoacylase 1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human Aminoacylase 1. This antibody recognizes human,mouse,rat antigen. The Aminoacylase 1 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

Type: Primary
Antigen: ACY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty