Would you like to log in again?
BAMBANKER™, Serum Free Cell Freezing Medium, Wako Chemicals
Supplier: Fujifilm Wako Chemicals USA
302-14681
101974-112EA 172.62 USD InStock
101974-112
BAMBANKER™, Serum Free Cell Freezing Medium, Wako Chemicals
Cell Freezing Media
Serum-free freezing medium for tumor and normal cells.
  • Ready-to-use medium for preservation of cells
  • Usable without diluting
  • Long-term freezing
BAMBANKER™ is for long-term freezing at -80 ºC (-112 ºF) and preservation of valuable culture cells such as tumor cells and normal cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty