Would you like to log in again?
Totally Tubular Tube Rack System, USA Scientific
Supplier: USA Scientific
2344-1000 2344-1000
102952-534EA 56.77 USD InStock
102952-534 82020-734
Totally Tubular Tube Rack System, USA Scientific
Racks Tube Racks
Totally Tubular modular rack system includes one set of four tube blocks in mixed standard colors and one tray per pack.
  • Four removable tube blocks
  • Made of durable polypropylene
Modular rack includes four removable blocks that can be positioned to support four sizes of tubes
Ideal for 15 and 50mL conical tubes; 1.5 and 2.0mL microcentrifuge tubes; and 12 x 75, 13 x 100, 16 x 125, and 17 x 100mm culture tubes.
Durable polypropylene withstands freezing and autoclaving. Mixed standard colors include blue, green, red, and yellow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty