Would you like to log in again?
GeneMate Combination Packs, 8-Strip Standard Tubes with 8-Strip Optically-Clear Flat Caps, Scientific Specialties
Supplier: Scientific Specialties
3135-00-210IS
490004-440BX 291.91 USD InStock
490004-440
GeneMate Combination Packs, 8-Strip Standard Tubes with 8-Strip Optically-Clear Flat Caps, Scientific Specialties
Tubes PCR Tubes
Designed to precisely fit most thermal cyclers. Ideal for qPCR (real-time PCR). Convenient tube and cap combination packs. Equal number of tubes and caps in one pack.
  • Secure evaporation-resistant seal between cap lid and tube
  • Optically-clear flat caps
  • Fluorescent signal is passed through unaffected
  • Ultra-thin wall for optimum heat transfer and higher yield
  • Tubes are pre-numbered for easy well identification
  • Crystal-clear clarity, easy to visualize content of each well
  • Available in white
Ideal for enhancing signal strength for qPCR assays.
Certified free DNase, RNase, and detectable DNA.
Ordering information: GeneMate 96-well PCR tube storage racks are available separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty