Would you like to log in again?
Rappaport-Vassiliadis R10 Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
K167
10789-322PK 68.27 USD InStock
10789-322
Rappaport-Vassiliadis R10 Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective enrichment of Salmonella spp
from meat, feces and water.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty