Would you like to log in again?
AK #2 Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
U189 U189
10789-324PK 547.65 USD InStock
10789-324 CA10789-324
AK #2 Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For preparation of spore suspensions used to detect antibiotic residues (such as penicillin and sulfa drugs) in milk and dairy products.
Ordering information: This product has a minimum order requirement of 3 packs of 12.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty