Would you like to log in again?
Salt Bridge
Supplier: EISCO SCIENTIFIC LLC
CH0744
470163-928EA 36.7 USD InStock
470163-928
Salt Bridge
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Comprises an inverted “U” tube with its ends closed by sintered discs. For use in experiments involving the measurement of cell EMF under conditions of forward, zero and reverse current flow. The bridge comprises an inverted “U” tube with its ends closed by sintered discs. This apparatus provides a more effective method of making reversible cells then the wet filter paper or agar gel methods. Dia. of arms 13mm, overall width 80mm, overall height 100mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty