Would you like to log in again?
GeneMate Individual PCR Tubes, with Domed Cap, 0.2 ml, Scientific Specialties
Supplier: Scientific Specialties
3210-09-110IS 3210-00-110IS
490003-652BX 144.08 USD InStock
490003-652 490003-640
GeneMate Individual PCR Tubes, with Domed Cap, 0.2 ml, Scientific Specialties
Tubes PCR Tubes
Secure evaporation-resistant seal between cap lid and tube.
  • Ultra-thin wall for optimum heat transfer and higher yield
  • Crystal clear clarity allows easy viewing of contents for each well
Available in a full range of colors.
Certified free from DNase, RNase, and detectable DNA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty