Would you like to log in again?
GeneMate 12-Strip Optically-Clear Flat Caps, Scientific Specialties
Supplier: Scientific Specialties
3106-00-400IS
490003-558BX 183.7 USD InStock
490003-558
GeneMate 12-Strip Optically-Clear Flat Caps, Scientific Specialties
Closures Test/Sample Tube Closures
Secure evaporation-resistant seal for strip tubes. Ideal for qPCR (real-time PCR).
  • Optically-clear
  • Fluorescent signal is passed through unaffected
  • Crystal-clear clarity
  • Easy to visualize contents through caps
  • Certified free from Dnase, Rnase, and detectable DNA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty