Would you like to log in again?
Preserved Rats
Supplier: Avantor
470158-756EA 20 USD InStock
470158-756 470158-754 470158-758 470158-760
Preserved Rats
Specimens Preserved Specimens Preserved Animal Materials
Formalin-preserved rats are available in vacuum packs.
  • Mammalian Anatomy
  • General Dissection
  • Formalin-Preserved Mink
  • In Vacuum Packs
  • Plain-Preserved, Single, Double, and Triple-Injected Rats
Our rat specimens are carefully preserved in an extended position, so students to examine their organ systems, and their skeletal, muscular, and reproductive structures.
Teachers can choose from plain preserved, single, double, and triple-injected rats. Injected rats are made without incisions in the neck, ensuring the specimens are intact.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty