Would you like to log in again?
Quantofix Aluminum Test Kit, CTL Scientific Supply
Supplier: CTL Scientific Supply
91307
10218-502EA 207.12 USD InStock
10218-502
Quantofix Aluminum Test Kit, CTL Scientific Supply
Environmental Test Kits
Quantofix Aluminum test strips and reagent for the semi-quantitative determination of aluminum in solutions.
Quantofix Aluminum has a gradation of scale: 0-5-20-50-200-500 mg/L (ppm) Al3+.
Ordering information: Packaged as a box of 100, strips and reagent.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty