Would you like to log in again?
GeneMate 12-Strip Domed Caps, Scientific Specialties
Supplier: Scientific Specialties
3103-00-400IS
490003-542BX 119.19 USD InStock
490003-542
GeneMate 12-Strip Domed Caps, Scientific Specialties
Closures Test/Sample Tube Closures
Secure evaporation-resistant seal for strip tubes.
  • Crystal-clear clarity
  • Easy to visualize content through caps
  • Certified free from Dnase, Rnase, and detectable DNA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty