Would you like to log in again?
Balance Pan Liners, Aluminum, Dyn-a-Med
Supplier: Dyn-A-Med Products
80062 80040
11300-350CS 533.13 USD InStock
11300-350 11300-442
Balance Pan Liners, Aluminum, Dyn-a-Med
Boats Weighing Boats
Compatible generic aluminum moisture pans for most moisture balances.
  • Strong and lightweight
Multiple uses with liquid and solid samples including weighing, drying and moisture determination. The inexpensive disposable aluminum foil dishes with crimped sides and a flat smooth bottom are great for moisture testing, wastewater and many other applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty