Would you like to log in again?
Energy Transfer Apparatus
Supplier: EISCO SCIENTIFIC LLC
PH0467B
470163-046EA 31.25 USD InStock
470163-046
Energy Transfer Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
Demonstrate the conversion from electrical energy to mechanical energy
  • Investigate electrical to mechanical energy
Demonstrate the conversion of energy with this small motor and its' attached pulley. The unit can also be used with a larger motor/generator to compare power input and output. Drive each in turn with the other while metering the voltage and current.
Motor generator and hanging masses sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty