Would you like to log in again?
Chicken Serum, Vector Laboratories
Supplier: Vector Laboratories
S-3000-20
101098-386EA 198 USD InStock
101098-386
Chicken Serum, Vector Laboratories
Sera (Serum)
Normal chicken serum can be used as a blocking reagent in Immunoassays.
  • It is a pooled sample collected from healthy adult animals
  • Each serum is tested with the appropriate antibody to check for possible cross-reactivities
  • The sera are supplied undiluted with 0.08% sodium azide as a preservative
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty