Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Whiteboard Color Optics
Supplier: United Scientific Supplies
WBOPS1-220V WBOPS1 WBOPS1-W WBOPS1-A
470339-764EA 243.35 USD InStock
470339-764 470144-223 470218-758 470339-518
Whiteboard Color Optics
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Whiteboard Color Optics are a complete system for investigating color, reflection, and refraction.
  • Magnetic-Backed Components
  • Lenses and Color Filters Included
  • Instruction Manual Included
  • Halogen Lamp Included
  • Whiteboard Features Swiveling Feet
The set of equipment for ray optics and color experiments uses magnetic-backed components. It can be used on the included whiteboard or any installed steel whiteboard. The whiteboard has a usable surface of 23 x 15.5" and has swiveling feet on two sides, so it can be set up in wide or tall format.
The double-ended light box contains a 12V/20W halogen lamp, powered by a wall-mounted AC adapter and cooled by a built-in fan. One end carries a cylindrical lens and slots for mounting the three slit diaphragms for ray optics. The other end carries two adjustable mirrors and an open area for color mixing work.
Ordering information: This set includes one 23 x 15½” whiteboard, 1 double-ended light box, 1 convex acrylic lens, 1 concave acrylic lens, 1 semicircular acrylic lens, 1 rectangular acrylic block, 1 acrylic right angle prism, 1 acrylic equilateral prism, one 30°/60°/90° prism, 1 hollow acrylic rectangular cell for water refraction, 1 plane mirror, 1 adjustable concave/convex mirror, 1 matched set of seven color filters, 1 set of seven corresponding colored paddles, one 10 x 10” white card screen, 1 card protractor circle, 4 magnets to hold protractor, 1 experiment manual, and 1 storage box with molded inserts.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty