Would you like to log in again?
Fundamentals of Electricity
Supplier: MATRIX TECHNOLOGY SOLUTIONS, LTD
LK6444A
470163-890KT 341.95 USD InStock
470163-890
Fundamentals of Electricity
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This kit provides an introduction to the fundamentals of electricity.
  • Topics Include Two-Way Switches, Buzzers, and More
  • Carriers, Baseboard, and Power Supply Provided
  • Free Curriculum Provided
  • CD ROM Provided
  • For Ages Eight and Older
It's ideal for those who are new to the subject and is suitable for use from ages eight and up. The kit includes carriers, a baseboard, power supply, and 30 pages of notes and worksheets on CD ROM.
Learning objectives and experiments include electricity basics, simple electrical components, the simplest circuit, conductors and insulators, switches, two-way switches, series circuits, parallel circuits, buzzers, and motors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty