Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BOLL Goat Polyclonal Antibody

Supplier: Abnova
PAB6469
10691-432EA 527.69 USD InStock
10691-432
Anti-BOLL Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of BOLL.

Recommended Dilutions: ELISA: 1:32000

Type: Primary
Antigen: BOLL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty