Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCAR1 Mouse Monoclonal Antibody [clone: M144]

Supplier: Abnova

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

MAB1386
10690-860EA 527.69 USD InStock
10690-860
Anti-BCAR1 Mouse Monoclonal Antibody [clone: M144]
Antibodies
Mouse monoclonal antibody raised against partial recombinant Bcar1.

Recommended Dilutions: ELISA: 1:2000; Immunocytochemistry: 1:250; Immunoprecipitation: 5-10 ?l; Western Blot: 1:1000

Type: Primary
Antigen: BCAR1
Clonality: Monoclonal
Clone: M144
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty