Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCL2L1 Mouse Monoclonal Antibody [clone: 2H12]

Supplier: Abnova

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

MAB1802
10690-966EA 527.69 USD InStock
10690-966
Anti-BCL2L1 Mouse Monoclonal Antibody [clone: 2H12]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of BCL2L1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: BCL2L1
Clonality: Monoclonal
Clone: 2H12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty