Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-His Tag Mouse Monoclonal Antibody [clone: BHISH8]

Supplier: Biosensis
M-1311-100
10782-048EA 357 USD InStock
10782-048
Anti-His Tag Mouse Monoclonal Antibody [clone: BHISH8]
Antibodies
The 6X His tag is a short peptide sequence of 6 histidine residues. Epitopes such as the 6X His tag are often included with the target DNA at the time of cloning to produce fusion proteins containing the tag sequence. This allows anti-epitope tag antibodies such as this one to serve as a universal detection reagent for any recombinant protein containing this tag. Anti-epitope antibodies are a useful alternative to generating antibodies to identify a specific recombinant protein. The 6X His motif is often used as a tag on recombinant proteins to facilitate purification with immobilized metal-affinity chromatography.

His-tagged fusion proteins
Application Information:
Western Blotting (WB) and Immunoprecipitation (IP). Suggested starting dilutions are as follows: WB at dilutions of 1:1000, IP at dilutions of 1:200. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: His Tag
Clonality: Monoclonal
Clone: BHISH8
Conjugation: Unconjugated
Epitope: 6-His
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty