Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Beta Endorphin Rabbit Polyclonal Antibody

Supplier: Biosensis
R-1492-50
10782-486EA 327 USD InStock
10782-486
Anti-Beta Endorphin Rabbit Polyclonal Antibody
Antibodies
Human beta-endorphin is a 31 amino acid peptide cleaved from the precursor pro-opiomelanocortin (POMC). It is an endogenous opioid peptide neurotransmitter that interacts with opioid receptors.

Confirmed by RIA against the synthetic peptide.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. For radioimmunoassay (RIA), a working dilution of 1:10,000 is recommended. Sensitivity - 120 pg/sample; IC50-1 ng/ sample. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Beta Endorphin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty