Would you like to log in again?
Mist Eliminator
Supplier: FISCHER TECHNICAL CO SE
LAV-EF
470164-238EA 111.45 USD InStock
470164-238
Mist Eliminator
Pumps Vacuum Pumps
The pump exhaust filter eliminates smoke from the high vacuum pump exhaust by removing oil mists. 
  • Reduce noise levels
This will protect the working environment and maintain a retention efficiency of 99.9% at .1 um with low flow resistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty