Would you like to log in again?
Mouse Recombinant FGF2 (from E. coli)
Supplier: BioVision
AB285465-50UG AB285465-1MG AB285465-10UG
10190-184EA 268.12 USD InStock
10190-184 10190-186 10190-188
Mouse Recombinant FGF2 (from E. coli)
Proteins and Peptides
>95% Pure active Recombinant murine FGF-2, to stimulate the proliferation of a wide variety of cells including mesenchymal, neuroectodermal and endothelial cells.
The ED₅₀, calculated by the dose-dependant proliferation of BALB/3T3 cells is <0.5 ng/ml.
FGF-2 is a basic heparin binding growth factor that stimulates the proliferation of a wide variety of cells including mesenchymal, neuroectodermal and endothelial cells. The recombinant mouse FGF-2 is a single, non-glycosylated chain containing 145 amino acids and having a molecular weight of 16.3 kDa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty