Would you like to log in again?
Human Recombinant FGF21 (from E. coli)
Supplier: BioVision
AB285474-100UG AB285474-1MG AB285474-10UG
10190-214EA 536.26 USD InStock
10190-214 10190-216 10190-218
Human Recombinant FGF21 (from E. coli)
Proteins and Peptides
>95% Pure Recombinant Human FGF-21.
Recombinant Human FGF-21 produced in E.Coli is a single, non-glycosylated polypeptide chain containing 182 amino acids and having a molecular mass of 19.5 kDa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty