Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Wnt-1 Antibody

Supplier: BioVision
AB63934-100
10187-142EA 321.33 USD InStock
10187-142
Anti-Wnt-1 Antibody
Antibodies
Wnt gene family members, including Wnt-1 throμgh Wnt-10, play a key role in regulating cellular growth and differentiation. Wnt-1 is a cysteine-rich, secreted glycoprotein that associates with cell membranes and likely functions as a key regulator of cellular adhesion. Wnt-1, which is essential for normal development of the embryonic nervous system, contributes to hyperplasia and tumorigenic progression when improperly expressed in mammary tissue. Wnt-3 is also involved in tumorigenesis and Wnt-2 and Wnt-4 may be associated with abnormal proliferation in human breast tissue. Wnt-1, Wnt-3 and Wnt-10b have been implicated along with FGF-3 in the development of mouse mammary tumor virus induced mouse mammary carcinomas.

Western Blot (0.5 to 4 µg/ml), Immunohistochemistry (10 to 20 µg/ml). However, the optimal concentrations should be determined individually. The antibody recognizes 41kDa of human Wnt-1. Reactivity to other species has not been tested.

Type: Primary
Antigen: WNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty