Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ASAH2 Rabbit Polyclonal Antibody

Supplier: BioVision
AB63804-100
10187-174EA 321.33 USD InStock
10187-174
Anti-ASAH2 Rabbit Polyclonal Antibody
Antibodies
ASAH2 is a key regulator of sphingolipid signaling metabolites. It catalyzes the degradation of sphingolipid C-16 Ceramide into sphingosine and free fatty acid at an optimal pH of 6.5-8.5.

Western Blot (0.5-4 µg/ml). However, the optimal conditions should be determined individually. Other applications have not been determined.

Type: Primary
Antigen: ASAH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty