Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CERS5 Rabbit Polyclonal Antibody

Supplier: BioVision
AB63927-100
10187-178EA 321.24 USD InStock
10187-178
Anti-CERS5 Rabbit Polyclonal Antibody
Antibodies
Lass5 also known as longevity assurance homolog 5 is a dihydroceramide synthase. It uses palrnitoyl-CoA as acyl donor and is involved in the synthesis of C14, C16 and C18-ceramide.

Western Blot (0.5-4 µg/ml). Multiple isoforms of Lass5 can be determined. Other applications have not been determined.

Type: Primary
Antigen: CERS5
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty