Would you like to log in again?
Human Recombinant IL3 (from E. coli)
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB285507-1MG AB285507-50UG AB285507-10UG
10190-250EA 496.25 USD InStock
10190-250 10190-252 10190-254
Human Recombinant IL3 (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant Human IL-3, which stimulates colony formation of megakaryocytes, neutrophils, and macrophages from bone marrow cultures.
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is < 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10⁷ units/mg
IL-3 is a species-specific colony-stimulating factor that stimulates colony formation of megakaryocytes, neutrophils, and macrophages from bone marrow cultures. Recombinant human IL-3 is a 15.0 kDa protein containing 133 amino acid residues
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty