Would you like to log in again?
Mouse Recombinant IL3 (from E. coli)
Supplier: BioVision
AB285510-50UG AB285510-10UG AB285510-1000UG
10190-258EA 13230.23 USD InStock
10190-258 10190-260 10190-256
Mouse Recombinant IL3 (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant murine IL-3, which stimulates colony formation of megakaryocytes, neutrophils, and macrophages from bone marrow cultures.
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of murine M-NFS-60 cells is <0.05 ng/ml, corresponding to a specific activity of ≥2 x 10⁷ units/mg.
Interleukin-3 (IL-3) is a species-specific colony-stimulating factor that stimulates colony formation of megakaryocytes, neutrophils, and macrophages from bone marrow cultures. Recombinant murine IL-3 is a 15.1 kDa protein containing 135 amino acid residues
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty