Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Premium Heavy-Duty Bosshead

Supplier: EISCO SCIENTIFIC LLC
PRBH03
470165-426EA 15 USD InStock
470165-426
Premium Heavy-Duty Bosshead
Clamps Bossheads
Thicker Than Normal 7 mm Threaded Clamping Screws

  • Thicker casting to support more load
  • Accepts up to 15 mm rods
  • High-strength, powder-coated alloy construction
  • All metal construction to carry heavy loads and prevent torque damage

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty