Would you like to log in again?
Innovating Science® Introduction to Chemical Equations
Supplier: ALDON CORP SE
Innovating Science®
IS2514 IS2514-SGL
470231-578KT 55.85 USD InStock
470231-578 470165-508
Innovating Science® Introduction to Chemical Equations
Educational Classroom Kits and Activities
What happens when a chemical reaction takes place?
  • Students perform experiments that highlights differences in chemical reactions
  • Single and double replacement reactions included
  • Aligned to NGSS practices
  • Performance expectations: MS-PS1-2,5; HS-PS1-1,7
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts: Patterns, energy and matter
  • Engineering practices: Developing and using models, analyzing and interpreting data
Chemical reactions take place all around us. Being able to identify reactions and communicate what is happening is vital to the scientific community. This kit will introduce two common types of chemical reactions: single replacement and double replacement. Students will perform a series of experiments highlighting the differences in these reactions and use what they learn to write balanced chemical reaction equations. Kit contains enough materials for 15 groups. Teacher’s manual and Student Study Guide copymasters are included.
Delivery information: Instructor's manual and SDS included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty