Would you like to log in again?
Coverage® Plus NPD, Disinfectant, STERIS®
Supplier: Steris
Coverage®
638708
21899-844CS 559.2 USD InStock
21899-844
Coverage® Plus NPD, Disinfectant, STERIS®
Cleaning Agents Disinfectants
Coverage Plus NPD® One-Step Cleaner Disinfectant a phosphate-free, one-step cleaner/disinfectant formulated for use on hard non-porous surfaces.
  • Effective against Influenza, Herpes simplex Type 1 and 2, HIV-1 (AIDS virus)
  • It has no perfumes or dyes
  • Formulation includes surfactants for surface cleaning while disinfecting.
  • Ideal for use in a contamination control rotation.
  • Complete lot traceability.
  • Each lot documented by Certificate of Analysis
This product consists of four quaternary ammonium compounds formulated with a compatible detergent system. It is proven effective in the presence of heavy soil (5% bovine serum) and hard water. Coverage Plus NPD disinfectant contains no perfumes or dyes that could interfere with critical research studies or manufacturing processes.
It delivers 850 ppm active germicidal quat for broad spectrum effectiveness, when diluted with water, up to 400 ppm hardness (AOAC hard water) and in the presence of 5 % blood serum. It is used for simultaneous cleaning and disinfecting of washable, hard, non-porous environmental surfaces such as floors, walls, and processing equipment. Other surfaces that may be cleaned include glass, chrome, stainless steel, porcelain tile, washable painted or varnished surfaces, plastics, as well resilient vinyl, linoleum, asphalt, rubber or terrazzo floors.
Use-dilution is 1:256.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty