Would you like to log in again?
Whatman™ 850-DS 8-Channel Filter Plates, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Whatman™
7707-3500 7707-3500 7707-3400 7707-3600 7707-3700 7707-3900 7707-3600 7707-3000 7707-3200 7707-3300 7707-3800 7707-3100 7700-3800 7707-3700 7707-3300 7707-3100 7707-3000 7707-3400
75845-148PK 713.73 USD InStock
75845-148 CA75927-822 CA75927-824 CA75845-148 75845-150 CA75845-152 75959-658 CA75845-150 75845-152 75959-656 10141-518 CA10141-528 76332-570 CA10141-526 10141-514 CA10141-524 CA10141-522 10141-512
Whatman™ 850-DS 8-Channel Filter Plates, Whatman products (Cytiva)
Microplates Filter Plates
Whatman 8-channel filter plates are specifically designed for use with the Agilent™ 850-DS Dissolution Sampling Station.
  • Automated processing of up to 8 samples simultaneously
  • Saves time
  • Eliminate errors associated with manual sampling
Developed in conjunction with Agilent™, Whatman 850-DS 8-channel filter plates increase productivity by allowing reliable alignment of the liquid path and reducing the risk of jamming or leaks that may occur with other dissolution sample preparation systems.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty