Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DiscoveryPak™ Stem Cell Fate Regulator Set II, Biovision

Supplier: BioVision
AB286223-1KIT
10191-880EA 2083.73 USD InStock
10191-880
DiscoveryPak™ Stem Cell Fate Regulator Set II, Biovision
Cell Culture Supplements and Additives
A convenient set of eight small molecule modulators useful in stem cell research. The eight modulators are 5 mg of BIX01294, 5 mg of CHIR99021, 5 mg of PD 184352, 2 mg of PD-0325901, 10 mg of RG 108, 1 mg of SB-431542, 500 µg of SU-5402, and 200 mg of valproic acid sodium.

  • Appearance: Lyophilized solids
  • Purity: ≥95% by HPLC
  • Solubility: DMSO
  • Storage conditions: –20 °C

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty