Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DiscoveryPak™ Stem Cell Fate Regulator Set IV, Biovision

Supplier: BioVision
AB286225-1KIT
10190-094EA 819.72 USD InStock
10190-094
DiscoveryPak™ Stem Cell Fate Regulator Set IV, Biovision
Cell Culture Supplements and Additives
A convenient set of three small molecules useful for promoting the survival of embryonic stem cells (ESCs). The molecules are 1 mg of pyrintegrin, 1 mg of thiazovivin and 1 mg of Y-27632, HCl.

  • Appearance: Lyophilized solids
  • Purity: ≥95% by HPLC
  • Solubility: DMSO
  • Storage conditions: –20 °C

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty