Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DiscoveryPak™ Stem Cell Fate Regulator Set V, Biovision

Supplier: BioVision
AB286226-1KIT
10191-384EA 957.61 USD InStock
10191-384
DiscoveryPak™ Stem Cell Fate Regulator Set V, Biovision
Cell Culture Supplements and Additives
A convenient set of five small molecules useful for generating human induced pluripotent stem cells (iPSC) from primary cells. The five molecules are 1 mg of A-83-01, 2 mg of PD-0325901, 5 mg of PS48, 1 g of sodium butyrate and 25 mg of tranylcypromine hemisulfate (parnate).

  • Appearance: Lyophilized solids
  • Purity: ≥95% by HPLC
  • Solubility: DMSO
  • Storage conditions: –20 °C

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty