Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

DiscoveryPak™ Stem Cell Fate Regulator Set VI, Biovision

Supplier: BioVision
AB286227-1KIT
10190-104EA 1409.6 USD InStock
10190-104
DiscoveryPak™ Stem Cell Fate Regulator Set VI, Biovision
Cell Culture Supplements and Additives
A convenient set of five small molecules useful for self-renewal in stem cells and induced pluripotent stem cells (iPSCs). The five molecules are 1 mg of A-83-01, 5 mg of CHIR99021, 1 mg of IQ-1, 2 mg of PD-0325901, and 500 µg of SU-5402.

  • Appearance: Lyophilized solids
  • Purity: ≥95% by HPLC
  • Storage conditions: –20 °C

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty