Would you like to log in again?
StemBoost™ YPAC Cocktail Set (1000X), Sterile-Filtered, BioVision
Supplier: BioVision
AB286230-5X1ML AB286231-1ML K871-1SET
10190-126EA 974.85 USD InStock
10190-126 10190-122 10190-124
StemBoost™ YPAC Cocktail Set (1000X), Sterile-Filtered, BioVision
Cell Culture Supplements and Additives
A convenient, sterile-filtered cocktail solution (1000X, in DMSO) containing four signaling inhibitors useful for maintaining the pluripotency of embryonic stem cells (ESCs).
Delivery information: It includes four products in the cocktail are: ROCK inhibitor (YPAC 10.0 mM), MEK inhibitor (PD0325901 1.0 mM), ALK5 kinase inhibitor (A83-01 0.5 mM) and GSK-3β inhibitor (CHIR99021 3.0 mM).
Caution: For research use only. Not for use in humans.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty